Alfa Aesar™ Strontium peroxide, 12.3% available oxygen

Manufacturer: Alfa Aesar™ 46310A7

Quantity 5kg

 View more versions of this product

Catalog No. AA46310A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-18-7
    Molecular Formula O2Sr
    Linear Formula SrO2
    Formula Weight 119.62
    MDL Number MFCD00084686
    UN Number UN1509
    Merck Index 14,8847
    Chemical Name or Material Strontium peroxide, 12.3% available oxygen

    Specifications

    Formula Weight 119.62
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1314-18-7
    Health Hazard 2 GHS H Statement
    H272-H318-H335-H315
    May intensify fire
    oxidizer.
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    TSCA TSCA
    Mesh Size −100 Mesh
    Physical Form Powder
    Sensitivity Moisture Sensitive
    Molecular Formula O2Sr
    UN Number UN1509
    Merck Index 14,8847
    Color Pale brown
    Melting Point 215°C (dec.)
    Quantity 5kg
    Linear Formula SrO2
    MDL Number MFCD00084686
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: STRONTIUM PEROXIDE
    Chemical Name or Material Strontium peroxide, 12.3% available oxygen

    GHS H Statement
    H272-H318-H335-H315
    May intensify fire
    oxidizer.
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit