Strontium, plasma standard solution, Sr 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 13874AP

Quantity 500mL

 View more versions of this product

Catalog No. AA13874AP

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula Sr(NO3)2 in 5% HNO3
    MDL Number MFCD00134060
    Chemical Name or Material Strontium, plasma standard solution, Sr 1000μg/mL

    Specifications

    Quantity 500mL
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 231-714-2
    Physical Form Liquid
    Appearance Liquid
    Molecular Formula Sr(NO3)2 in 5% HNO3
    MDL Number MFCD00134060
    Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Strontium nitrate: 0.24%; Water: 94.76%
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Strontium, plasma standard solution, Sr 1000μg/mL

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit