Alfa Aesar™ Strontium sulfide, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 14473-14

Quantity 25g

 View more versions of this product

Catalog No. AA1447314

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1314-96-1
    Formula Weight 119.68
    MDL Number MFCD00049560
    Merck Index 14,8849
    Chemical Name or Material Strontium sulfide

    Specifications

    Formula Weight 119.68
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for severa
    Odor Like rotten eggs
    Physical Form Powder
    Sensitivity Hygroscopic
    Quantity 25g
    Health Hazard 1 Warning
    Density 3.70g/mL
    TSCA TSCA
    Color Gray
    MDL Number MFCD00049560
    Merck Index 14,8849
    Melting Point >2000°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Storage Note 1
    • Ampouled under argon
    CAS 1314-96-1
    Chemical Name or Material Strontium sulfide
    Assay 99.995%, (metals basis)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for severa

    • Ampouled under argon

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit