Alfa Aesar™ Strontium titanium oxide, 99+% (metals basis)

Used in electronics and in electrical insulation

Manufacturer: Alfa Aesar™ 11399A1

Quantity 1kg

 View more versions of this product

Catalog No. AA11399A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12060-59-2
    Formula Weight 183.51
    MDL Number MFCD00049554
    Synonym Strontium titanate
    Chemical Name or Material Strontium titanium oxide

    Specifications

    Formula Weight 183.51
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00049554
    Quantity 1kg
    Solubility Information Insoluble in water and most solvents
    Density 4.810g/mL
    TSCA TSCA
    CAS 12060-59-2
    Synonym Strontium titanate
    Melting Point 2060°C
    Chemical Name or Material Strontium titanium oxide
    Assay ≥99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit