Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Alfa Aesar™

Click to view available options
Quantity:
1 ea.
5 ea.
12060-59-2
O3SrTi
183.484
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium titanate,strontium titanium oxide,unii-olh4i98373,o3ti.sr,strontium titanium oxide srtio3,strontium titanium trioxide,titanate tio32-, strontium 1:1,strontium titanate 100g,strontium dioxido oxo titanium,strontium titanate, powder
82899
strontium;dioxido(oxo)titanium
[O-][Ti](=O)[O-].[Sr+2]
12060-59-2
O3SrTi
183.484
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium titanate,strontium titanium oxide,unii-olh4i98373,o3ti.sr,strontium titanium oxide srtio3,strontium titanium trioxide,titanate tio32-, strontium 1:1,strontium titanate 100g,strontium dioxido oxo titanium,strontium titanate, powder
82899
strontium;dioxido(oxo)titanium
[O-][Ti](=O)[O-].[Sr+2]
Solubility Information Insoluble in water.
Physical Form Substrate
Odor Odorless
Color White
Quantity 1 ea.
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side

EINECSNumber : 235-044-1

TSCA : Yes

Recommended Storage : Ambient temperatures