Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 110 Orientation, Thermo Scientific™

Catalog Number AA40278KT
Click to view available options
Quantity:
1 Ea.
5 Ea.
12060-59-2
O3SrTi
183.48
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
82899
strontium;dioxido(oxo)titanium
[Sr++].[O-][Ti]([O-])=O
12060-59-2
O3SrTi
183.48
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
82899
strontium;dioxido(oxo)titanium
[Sr++].[O-][Ti]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Substrate
Quantity 1 Ea.
Odor Odorless
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side
EINECSNumber 235-044-1
TSCA Yes
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit