Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished One Side, 110 Orientation, Alfa Aesar™

Manufacturer: Alfa Aesar™ 42619KU

Quantity 5 ea.

 View more versions of this product

Catalog No. AA42619KU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12060-59-2
    Molecular Formula SrTiO3
    MDL Number MFCD00049554
    Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side

    Specifications

    Physical Form Substrate
    Color White to off-white
    Quantity 5 ea.
    Odor Odorless
    Molecular Formula SrTiO3
    MDL Number MFCD00049554
    CAS 12060-59-2
    Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit