Learn More
Strontium Titanium Oxide Substrate, 5 x 5 x 1mm, Polished Two Sides, 100 Orientation, Alfa Aesar™
100 Orientation
Manufacturer:  Alfa Aesar 42670KU
Quantity 5 ea.
 View more versions of this product
Catalog No. AA42670KU
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
12060-59-2
SrTiO3
Strontium Titanium Oxide Substrate, Polished Two Sides
Substrate
MFCD00049554
Specifications
12060-59-2
SrTiO3
Odorless
5 ea.
Substrate
MFCD00049554
White to off-white
Strontium Titanium Oxide Substrate, Polished Two Sides