Alfa Aesar™ Strontium tungsten oxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4189914

Quantity 25g

 View more versions of this product

Catalog No. AA4189914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13451-05-3
    Linear Formula SrWO4
    Formula Weight 335.46
    MDL Number MFCD00049562
    Synonym Strontium tungstate
    Chemical Name or Material Strontium tungsten oxide

    Specifications

    Formula Weight 335.46
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White
    Physical Form Powder
    Linear Formula SrWO4
    CAS 13451-05-3
    Synonym Strontium tungstate
    Density 6.187g/mL
    TSCA TSCA
    Mesh Size −200 Mesh
    Odor Odorless
    MDL Number MFCD00049562
    Health Hazard 2 GHS H Statement
    H335
    May cause respiratory irritation.
    Quantity 25g
    Health Hazard 1 Warning
    Chemical Name or Material Strontium tungsten oxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H335
    May cause respiratory irritation.

    GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit