Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Stuart Analog Hot Plates

Custom designed hot plates provide accurate temperature control

 View more versions of this product

Catalog No. 11-475-026


Add to cart

Description

Description

  • Hot surface warning light flashes when plate temperature is above 50°C
  • Three color choices: blue, white, and black
Specifications

Specifications

Stuart analog hot plate
US-Style
1
Ceramic
450°C Max
Square
120 V
Black
6 x 6 in. plate
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit