Learn More
Dynalon™ Student Project Tanks
SDPSCI_ED
Student Project Tanks constructed of clear polycarbonate are designed to not crack or break.
Supplier:  Dynalon™ 4264540008
 View more versions of this product
Catalog No. S30765
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Molded of a shatter-resistant polycarbonate material that resists scratches, dents, and clouding-up
  • For long term classroom use
  • Tanks and lids also naturally resist many acids and oils to maintain a clear student view
  • Easy to clean and maintain
  • Can withstand the abuse of daily classroom use
  • Includes: 1 PC clear tank 1 PC clear lid, 1 PC clear drain shelf, 2 solid polyamide plugs, 2 vented polyamide plugs, 2 clear plastic/SS clamps
  • Pop-out plugs are supplied both as solid and vented allowing versatility in use
  • Included clamps will secure down the lid
  • Tank accessory pack includes 2 of each plug and 2 plastic clips
Specifications
Specifications
3 gal.
Polycarbonate
Education
12.75 in.
2.8 lb.
11.35 L
Aquarium/Terrarium Ventilated Cover
1/Ea.
323.85 mm