Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STX17 Mouse anti-Human, Biotin, Clone: 12610, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243796B

Catalog No. NBP243796B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

STX17
12610
Western Blot
Purified
STX17
Recombinant protein encompassing a sequence within the center region of human STX17. The exact sequence is proprietary.
Protein A purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Biotin
PBS with 0.05% Sodium Azide
MGC102796, MGC126613, MGC126615, syntaxin 17, syntaxin-17
Mouse
IgG2a
0.1 ml
Primary
55014
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit