Learn More
STX17 Mouse anti-Human, Clone: GT1252, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527839
Catalog No. PIMA527839
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Positive Control: HeLa, HeLa nuclear, 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (96%), Zebrafish (92%), Drosophila (88%), Yeast (84%), Xenopus laevis (92%), Chicken (91%), Rhesus Monkey (100%), Bo
STX17 encodes a product that has been implicated in vesicle trafficking to lysosomes. Could be involved in processes related to cell division.
Specifications
Specifications
STX17
Monoclonal
1.9 mg/ml
PBS with no preservative
P56962
syntaxin 17
Mouse
IgG2a
100 μl
-20° C, Avoid Freeze/Thaw Cycles
Primary
55014
Western Blot
GT1252
Unconjugated
STX17
Liquid
STX17
Recombit protein encompassing a sequence within the center region of human STX17. The exact sequence is proprietary.
Protein A
RUO
Antibody
Monoclonal
Human