Learn More
STX18 Mouse anti-Human, Clone: OTI2C4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809376
Catalog No. 50-166-9458
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
Specifications
Specifications
STX18
Monoclonal
Unconjugated
STX18
Lyophilized
STX18
Human recombit protein fragment corresponding to amino acids 1-285 of human STX18 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2C4
PBS with 8% trehalose and no preservative; pH 7.3
Q9P2W9
Ufe1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
53407