Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STX19 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548216

Catalog No. PA548216

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

STX19
Polyclonal
PBS with 0.09% sodium azide
Q8N4C7, Q8R1Q0
Syntaxin-19, STX19
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
415117, 68159
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
STX19
Liquid
STX19
KLH conjugated synthetic peptide between 250-279 amino acids from the C-terminal region of human STX19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit