Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stxbp5l Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$153.04 - $436.00
Specifications
Antigen | Stxbp5l |
---|---|
Concentration | LYOPH |
Dilution | Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10091288
|
Novus Biologicals
NB10091288 |
0.5 mg |
Each for $436.00
|
|
NB017896
|
Novus Biologicals
NB100912880.1MG |
0.1 mg |
Each for $153.04
|
|
Description
Stxbp5l Polyclonal specifically detects Stxbp5l in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Stxbp5l | |
Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
KIAA1006, lethal(2) giant larvae protein homolog 4, syntaxin binding protein 5-like | |
STXBP5L | |
IgG | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse | |
Q5DQR4 | |
9515 | |
A synthetic peptide as a part of mouse Syntaxin-binding protein 5-like, isoforms 1 to 5 (Tomosyn-2, Stxbp5l, Llgl4, STB5L) conjugated to an immunogenic carrier protein was used as the immunogen. | |
Primary | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
Stxbp5l. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title