Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STYK1 Antibody (484713), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6668T
Description
STYK1 Monoclonal antibody specifically detects STYK1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
STYK1 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
484713 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
DKFZp761P1010, EC 2.7.10.2, NOKProtein PK-unique, serine/threonine/tyrosine kinase 1Novel oncogene with kinase domain, SuRTK106, tyrosine-protein kinase STYK1 | |
E. coli-derived recombinant human STYK1, Asp72-Val320, Accession # Q6J9G0 | |
0.1 mL | |
Protein Kinase | |
55359 | |
Store at 4C in the dark. | |
IgG2a |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction