Alfa Aesar™ Styrene, 99.5% stab. with 4-tert-butylcatechol

Manufacturer: Alfa Aesar™ 16199K2

Quantity 1L

 View more versions of this product

Catalog No. AA16199K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-42-5
    Formula Weight 104.15
    MDL Number MFCD00008612
    UN Number UN2055
    Beilstein 1071236
    Merck Index 14,8860
    Synonym Phenylethylene; Vinylbenzene
    Chemical Name or Material Styrene, Stabilized with 4-tert-butylcatechol
    Grade Technical Grade

    Specifications

    Formula Weight 104.15
    Health Hazard 2 GHS H Statement
    H372-H304-H361-H226-H332-H315-H319-H335
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.906g/mL
    Physical Form Liquid
    UN Number UN2055
    Melting Point −31°C
    Synonym Phenylethylene; Vinylbenzene
    Health Hazard 1 Danger
    Storage Note 1 2°C to 8°C
    pH neutral
    Flash Point 31° (88°F)
    Odor Pungent
    MDL Number MFCD00008612
    CAS 100-42-5
    Beilstein 1071236
    Boiling Point 145° to 146°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED
    TSCA TSCA
    Merck Index 14,8860
    Refractive Index 1.5460
    Color Colorless
    Quantity 1L
    RTECS Number WL3675000
    Sensitivity Light Sensitive
    Chemical Name or Material Styrene, Stabilized with 4-tert-butylcatechol
    Assay 99.5%
    Grade Technical Grade

    GHS H Statement
    H372-H304-H361-H226-H332-H315-H319-H335
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2°C to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit