Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrene Monomer (Stabilized/Certified), Fisher Chemical

Click to view available options
Quantity:
1L
Packaging:
Glass Bottle
100-42-5
C8H8
104.15
MFCD00008612,MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
ethenylbenzene
C=CC1=CC=CC=C1
100-42-5
C8H8
104.15
MFCD00008612,MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
ethenylbenzene
C=CC1=CC=CC=C1
Vapor Pressure 7mbar at 20°C
Packaging Glass Bottle
Quantity 1L
Vapor Density 1.22g/mol
Refractive Index 1.545 to 1.547 (at 20°C)
Identification Pass Test
Boiling Point 145°C
Melting Point -31°C
Color Colorless
Density 0.906g/cm³
Linear Formula C6H5CH:CH2
Formula Weight 104.15
Physical Form Liquid
Percent Purity ≥99%
Grade Certified
Chemical Name or Material Styrene Monomer, Stabilized
Show More Show Less