Learn More
Styrofoam Balls
SCI_ED
Low-cost alternative to wood molecular model kits.
$48.75
Specifications
Diameter (English) 2 in.
Diameter (Metric) 50mm
Type Styrofoam balls
View More Specs
Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
S44062CBK
SCI_ED
View Documents Promotion Details
Denoyer Geppert Company
FOM304
Black
Pack of 25 for $48.75
Add to cart
S44062CBL
SCI_ED
View Documents Promotion Details
Denoyer Geppert Company
FOM307
Blue
Pack of 25 for $48.75
Add to cart
S44062CGR
SCI_ED
View Documents Promotion Details
Denoyer Geppert Company
FOM306
Green
Pack of 25 for $48.75
Add to cart
S44062CRD
SCI_ED
View Documents Promotion Details
Denoyer Geppert Company
FOM305
Red
Pack of 25 for $48.75
Add to cart
S44062CYW
SCI_ED
View Documents Promotion Details
Denoyer Geppert Company
FOM308
Yellow
Pack of 25 for $48.75
Add to cart
Description
Description
  • Build styrofoam molecules with pipe cleaner bonds
  • Flexible pipe cleaners pushed through the balls hold the model together
  • White styrofoam balls in a variety of sizes to differentiate between elements
  • To further differentiate, dip balls into water-soluble paint
  • Also available, 50mm styrofoam balls in a variety of colors
Specifications
Specifications
2 in.
50mm
Styrofoam balls