Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrylboronic acid, 97%

Click to view available options
Quantity:
500mg
2.5g
Packaging:
Glass bottle
6783-05-7
C8H9BO2
147.97
VKIJXFIYBAYHOE-VOTSOKGWSA-N
styrylboronic acid,e-styrylboronic acid,e-phenylethenylboronic acid,trans-2-phenylvinylboronic acid,e-2-phenylethenyl boronic acid,2-phenylvinylboronic acid,trans-phenylethenylboronic acid,trans-phenylvinylboronic acid,e-styreneboronic acid
5702628
[(E)-2-phenylethenyl]boronic acid
B(C=CC1=CC=CC=C1)(O)O
6783-05-7
C8H9BO2
147.97
VKIJXFIYBAYHOE-VOTSOKGWSA-N
styrylboronic acid,e-styrylboronic acid,e-phenylethenylboronic acid,trans-2-phenylvinylboronic acid,e-2-phenylethenyl boronic acid,2-phenylvinylboronic acid,trans-phenylethenylboronic acid,trans-phenylvinylboronic acid,e-styreneboronic acid
5702628
[(E)-2-phenylethenyl]boronic acid
B(C=CC1=CC=CC=C1)(O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow to Beige
Quantity 2.5g
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 147.97
Physical Form Powder and/or Chunks
Percent Purity ≥94% (HPLC)
Chemical Name or Material trans-β-Styreneboronic acid
Show More Show Less

GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

Warning