Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suberic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
505-48-6
C8H14O4
174.2
MFCD00004428
TYFQFVWCELRYAO-UHFFFAOYSA-N
suberic acid,cork acid,1,6-hexanedicarboxylic acid,1,8-octanedioic acid,hexamethylenedicarboxylic acid,subericacid,1,6-dicarboxyhexane,octane-1,8-dioic acid,octanedioc acid,unii-6u7y4m9c1h
10457
CHEBI:9300
octanedioic acid
C(CCCC(=O)O)CCC(=O)O
505-48-6
C8H14O4
174.2
MFCD00004428
TYFQFVWCELRYAO-UHFFFAOYSA-N
suberic acid,cork acid,1,6-hexanedicarboxylic acid,1,8-octanedioic acid,hexamethylenedicarboxylic acid,subericacid,1,6-dicarboxyhexane,octane-1,8-dioic acid,octanedioc acid,unii-6u7y4m9c1h
10457
CHEBI:9300
octanedioic acid
C(CCCC(=O)O)CCC(=O)O
Flash Point 203°C
Boiling Point 230°C (15.0mmHg)
Viscosity 6.2 mPa.s (150°C)
Melting Point 141°C to 144°C
Quantity 2.5kg
Packaging Plastic bottle
Infrared Spectrum Authentic
Color White to Cream
Assay Percent Range 99%
Linear Formula HO2C(CH2)6CO2H
Beilstein 02, IV, 2028
Merck Index 15, 8992
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: 1g/172mL ether,soluble in alcohol,practically insoluble in chloroform,soluble in polar solvents
Formula Weight 174.2
Physical Form Powder
Percent Purity 98.5 to 101.5%
Chemical Name or Material Suberic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 208-010-9

TSCA : TSCA