Alfa Aesar™ Suberoyl chloride, 97%

Manufacturer: Alfa Aesar™ B2458014

Quantity 25g

 View more versions of this product

Catalog No. AAB2458014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10027-07-3
    Molecular Formula C8H12Cl2O2
    Linear Formula ClOC(CH2)6COCl
    Formula Weight 211.09
    MDL Number MFCD00000764
    UN Number UN3265
    Synonym Octanedioyl chloride; Suberyl chloride
    Chemical Name or Material Suberoyl chloride

    Specifications

    Formula Weight 211.09
    Molecular Formula C8H12Cl2O2
    Linear Formula ClOC(CH2)6COCl
    MDL Number MFCD00000764
    CAS 10027-07-3
    UN Number UN3265
    Synonym Octanedioyl chloride; Suberyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 162° to 163°C (15mmHg)
    Flash Point >110°C (230°F)
    Quantity 25g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Suberoyl chloride
    Assay 97%

    Description

    Applications
    Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

    Solubility
    Reacts with water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit