Learn More
Koehler™ Bright Star™ Submersible Responder™ Flashlights
UL listed for use in hazardous locations, these watertight flashlights are backed by a limited lifetime warranty
Supplier: Koehler™ Bright Star™ 19102

Description
Choose the style that best suits your application.
C-Cell Flashlight
UL listed for use in hazardous locations classified as Class I, Division 1, Group D Temperature Rating T3A; Class I, Division 2, Groups A, B, C and D, Temperature Rating T3; and Class II, Division 2, Group G Temperature Rating T3.
- Traditional baton-style
- Features a push button switch and prefocused beam
- Impact-resistant design survives falls from as high as 30 feet; submersible to 100 feet
- Maximum candle power: 50,000 CP
- Requires 3 “C”-cell batteries
- Length: 8 3/4 in. (21.9cm)
- Yellow
UL listed for use in hazardous locations rated Class I, Division 1, Groups C and D Temperature Rating T3C; Class I, Division 2, Group A, B, C, and D Temperature Rating T3C; and Class II, Division 2, Group G Temperature Rating T3C.
- Small but effective flashlight fits into helmet clip
- Xenon bulb provides 13,000 CP
- Super tough nylon body survives fall from 30 feet and is submersible to 100 feet
- With push button switch and prefocused spot beam
- Length: 6 3/4 in. (17.1cm)
- Black, yellow, or orange
UL listed for use in hazardous locations classified as Class I, Division 2, Groups A, B, C, and D Temperature Rating T5, and Class II, Division 2, Group G Temperature Rating T5.
- Compact size and bright 5,000 CP beam makes this light exceptionally handy
- Attach to your key chain or keep it tucked away in your pocket
- Useful in tight or awkward spots such as computer repair or map reading
- Weighs less than an ounce without battery
- Submersible to 500 feet
- Length: 2 3/4 in. (7cm)
- Black or yellow
Submersible to 500 feet, this model is UL listed for use in locations classified as Class I, Division 1, Group C and D, Temperature Rating T3C; Class I, Division 2, Groups A, B, C and D, Temperature Rating T3C; and Class II, Division 2, Group G, Temperature Rating T3C.
- Purchase light individually or packaged with toolbox containing lightbender, red lens, replacement bulb module, O-ring, and key ring
- Length: 4 1/2 in. (11.4cm)
Specifications
Flashlight | |
15 hr. | |
0.46 kg | |
C Alkaline | |
UL, MSHA | |
Push-button | |
5.8 cm | |
8.62 in. | |
Polycarbonate |
Incandescent | |
16.3 oz. | |
5 hr. | |
50,000 cd | |
Yellow | |
2.31 in. | |
For use in hazardous locations, these watertight flashlights are backed by a limited lifetime warranty | |
21.8 cm | |
Nylon |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.