Learn More
Enzo Life SciencesSupplier Diversity Partner Substance P ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900018
Highly sensitive Substance P ELISA kit enabling detection of Substance P in plasma, serum, saliva, urine, and supernatants in just 3 hours. The Substance P EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit utilized low sample volume and provides the ability to assay up to 38 samples in duplicate per 96 well kit. Sensitivity: 8.04 pg/ml (range 9.76 - 10,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-104