Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Substance P Guinea Pig anti-Human, Mouse, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$231.88 - $506.30

Specifications

Antigen Substance P
Concentration Conc. Not Determined
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0007 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP14103150UL
150 μL
Each for $506.30
Add to cart
 
50-210-0008 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1410350UL
50 μL
Each for $231.88
Add to cart
 
Description

Description

Substance P Polyclonal antibody specifically detects Substance P in Human, Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

Substance P
Immunohistochemistry
Unconjugated
Guinea Pig
Human, Mouse, Rat
P06767, P20366, P41539
21333, 24806, 6863
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
4930528L02Rik; C-terminal-flanking peptide; Hs.2563; MGC26719; neurokinin 1; neurokinin 2; Neurokinin A; neurokinin alpha; Neuromedin L; Neuropeptide gamma; Neuropeptide K; NK1; NK-1; NK2; NKA; Nkna; NPK; PPT; Ppt5fl; PPTA; PPT-A; PPTA3; precursor 1; preprotachykinin; preprotachykinin A; protachykinin; Protachykinin-1; RAT TAC; RATPPTA3; SP; Substance K; Substance P; TAC; TAC1; Tac2; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
NH2-CRPKPQQFFGLM-CONH2-Corresponding to residues 1-11 of Rat Substance P
Polyclonal
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
TAC1
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit