Learn More
Bachem Z-Gly-Phe-OH, 25g, 1170-76-9, MFCD00037264 DFS Item
Manufacturer:  Bachem 4000159.0025
Substrate for carboxypeptidase A. The peptide is also an effective deactivating carrier into cells for primary and secondary amines, which are then unmasked intracellularly by enzymatic action.
Catalog No. 50-194-7013