Learn More
Bachem Suc-Ala-Ala-Phe-AMC, 250mg, 71973-79-0, MFCD00038448 DFS Item
Manufacturer:  Bachem 4006306.0250
Suc-AAF-AMC, a highly sensitive fluorogenic chymotrypsin substrate. It is also hydrolyzed by neutral metalloendopeptidases, such as thermolysin and enkephalinase (neprilysin, NEP 24.11). Suc-AAF-MCA has also been used for the chymotrypsin-like activity of the 20S proteasome.
Catalog No. 50-194-7030