Learn More
Peptides InternationalSupplier Diversity Partner Suc-Phe-Ala-Ala-Phe-pNA, 1mg, CAS# 110906-89-3, MF= C34H38N6O9 DFS Item
Manufacturer:  Peptides InternationalSupplier Diversity Partner SUB3007PI50MG
Application: Subtilisin and Myroilysin Substrate, salt: Trifluoroacetate Salt, one letter code: Suc-FAAF-pNA, MW: 674.72
Catalog No. 50-195-2436