Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Succinic Acid, ≥99.5% (T), Honeywell™ Fluka™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
110-15-6
C4H6O4
118.088
MFCD00002789
KDYFGRWQOYBRFD-UHFFFAOYSA-N
succinic acid,amber acid,asuccin,dihydrofumaric acid,bernsteinsaure,katasuccin,wormwood acid,succinate,ethylenesuccinic acid,1,2-ethanedicarboxylic acid
1110
CHEBI:15741
butanedioic acid
C(CC(=O)O)C(=O)O
110-15-6
C4H6O4
118.088
MFCD00002789
KDYFGRWQOYBRFD-UHFFFAOYSA-N
succinic acid,amber acid,asuccin,dihydrofumaric acid,bernsteinsaure,katasuccin,wormwood acid,succinate,ethylenesuccinic acid,1,2-ethanedicarboxylic acid
1110
CHEBI:15741
butanedioic acid
C(CC(=O)O)C(=O)O

Matrix substance for MALDI-MS,≥99.5% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Additional Information Matrix substance for MALDI-MS
Linear Formula HOOCCH2CH2COOH
UN Number NONH for all modes of transport
Beilstein 1754069
Density 1.56g/cu.cm
Formula Weight 118.09g/mol
Percent Purity ≥99.5% (T)
Packaging Glass Bottle
Solubility Soluble in methanol, 0.4g/10 mL
Physical State Crystals
Melting Point 184°C to 186°C
pH 2.7 (10.00g/L at 20°C)
Color White
Boiling Point 235°C
Quantity 5g
Flash Point 206°C
Chemical Name or Material Succinic Acid
Show More Show Less

P280-P305 + P351 + P338

H318

EINECSNumber : 203-740-4

RTECSNumber : WM4900000