Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Succinic Acid ACS AR Granular, Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
Specifications
Formula Weight | 118.09 |
Grade | ACS |
Quantity | 2.5 kg |
Packaging | Poly |
Physical Form | Granular |
Chemical Name or Material | Succinic Acid |