Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Succinic Acid, BAKER ANALYZED™ ACS Reagent, J.T. Baker™
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Description
- For Laboratory,Research,or Manufacturing Use
Specifications
Melting Point | 185.0°C to 191.0°C |
Formula Weight | 118.09 |
Grade | Meets ACS reagent chemical requirements |
Quantity | 2.5 kg |
Packaging | Poly Bottle |
Physical Form | Powder |
Specific Gravity | 1L = 1.57kg |
Chemical Name or Material | Succinic Acid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction