Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, Disodium Salt, 6-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
250g
5kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

6106-21-4
C4H16Na2O10
270.142
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
3083938
CHEBI:63686
disodium;butanedioate;hexahydrate
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 270.15
Packaging Poly Bottle
Quantity 250g
Chemical Name or Material Succinic Acid, Disodium Salt, 6-Hydrate
Grade BAKER ANALYZED™ Reagent