Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Succinic acid, Free acid, ≥99%, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
110-15-6
C4H6O4
118.088
MFCD0000278
KDYFGRWQOYBRFD-UHFFFAOYSA-N
succinic acid,amber acid,asuccin,dihydrofumaric acid,bernsteinsaure,katasuccin,wormwood acid,succinate,ethylenesuccinic acid,1,2-ethanedicarboxylic acid
1110
CHEBI:15741
butanedioic acid
C(CC(=O)O)C(=O)O
110-15-6
C4H6O4
118.088
MFCD0000278
KDYFGRWQOYBRFD-UHFFFAOYSA-N
succinic acid,amber acid,asuccin,dihydrofumaric acid,bernsteinsaure,katasuccin,wormwood acid,succinate,ethylenesuccinic acid,1,2-ethanedicarboxylic acid
1110
CHEBI:15741
butanedioic acid
C(CC(=O)O)C(=O)O
Linear Formula HOOCCH2CH2COOH
Formula Weight 118.09
Physical Form Crystalline Powder
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 1.56
Melting Point 186°C to 188°C
Boiling Point 235°C
Quantity 1kg
Flash Point >110°C (230°F)
Chemical Name or Material Succinic acid
Show More Show Less