Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Succinic acid, Free acid, ≥99%, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Specifications
Melting Point | 186°C to 188°C |
Boiling Point | 235°C |
Linear Formula | HOOCCH2CH2COOH |
Beilstein | 1754069 |
Formula Weight | 118.09 |
Percent Purity | ≥99% |
Density | 1.56 |
Quantity | 5 kg |
Flash Point | >110°C |
Packaging | Plastic bottle |
Show More |
Safety and Handling
Hazard Category | H318-H335 |
Precautionary Statement | P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c |
EINECSNumber | 203-740-4 |
RTECSNumber | WM4900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction