Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Succinic Acid, Ultrapure, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Succinic Acid, Ultrapure is a white, odorless solid that is a dicarboxylic acid. Advances in industrial biotechnology that seek to displace petroleum-based chemicals in industrial applications are fueling growth in this product. Spectrum’s Ultrapure grade products are suitable for use where the lowest levels of metals, organics, and particulates are required. These materials meet or exceed the specifications as listed in Reagent Chemicals published by the American Chemical Society.
Specifications
CAS Max % | 1 |
Percent Purity | 99% |
Grade | Ultrapure |
Quantity | 100 g |
Ignition Residue | 0.0002 |
Packaging | Poly Bottle |