Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, Ultrapure, 99%, Spectrum™

Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

S1983, 110-15-6, C4H6O4

Spectrum™ Succinic Acid, Ultrapure is a white, odorless solid that is a dicarboxylic acid. Advances in industrial biotechnology that seek to displace petroleum-based chemicals in industrial applications are fueling growth in this product. Spectrum’s Ultrapure grade products are suitable for use where the lowest levels of metals, organics, and particulates are required. These materials meet or exceed the specifications as listed in Reagent Chemicals published by the American Chemical Society.

CAS 110-15-6
CAS Max % 1
Calcium (Ca) 1e-005
Molecular Formula C4H6O4
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Copper (Cu) 5e-006
SMILES OC(=O)CCC(O)=O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
Percent Purity 99%
Grade Ultrapure
Ignition Residue 0.0002
Insoluble Matter 0.0001
Iron (Fe) 5 ppm
Lead (Pb) 1e-005
Magnesium (Mg) 5e-006
Packaging Poly Bottle
Potassium (K) 5e-005
Sodium (Na) 5e-005
Zinc (Zn) 5e-006
Quantity 500 g
Show More Show Less