Alfa Aesar™ Succinic anhydride, 99%

Manufacturer: Alfa Aesar™ A122450B

Quantity 1000g

 View more versions of this product

Catalog No. AAA122450B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-30-5
    Molecular Formula C4H4O3
    Formula Weight 100.07
    MDL Number MFCD00005525
    Beilstein 108441
    Merck Index 14,8870
    Synonym Butanedioic anhydride; Dihydro-2,5-furandione
    Chemical Name or Material Succinic anhydride

    Specifications

    Formula Weight 100.07
    Assay Percent Range 99%
    Storage Note 1 Moisture Sensitive
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Soluble in chloroform
    Beilstein 108441
    Flash Point 157°C (314°F)
    RTECS Number WN0875000
    Molecular Formula C4H4O3
    CAS 108-30-5
    Merck Index 14,8870
    Melting Point 118° to 121°C
    Quantity 1000g
    TSCA TSCA
    MDL Number MFCD00005525
    Synonym Butanedioic anhydride; Dihydro-2,5-furandione
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H319-H335
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 261°C
    Chemical Name or Material Succinic anhydride
    Assay 99%

    Description

    Applications
    Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

    Solubility
    Soluble in chloroform.Soluble in chloroform, carbon tetrachloride and alcohol. Slightly soluble in ether. Insoluble in water.

    Notes
    Moisture sensitive. Incompatible with strong oxidizing agents, alcohols and amines.

    GHS H Statement
    H302-H319-H335
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Moisture Sensitive

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit