Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinyl Chloride, ca. 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
543-20-4
C4H4Cl2O2
154.98
MFCD00000749
IRXBNHGNHKNOJI-UHFFFAOYSA-N
succinyl chloride,succinyl dichloride,succinic acid dichloride,succinoyl chloride,succinoyl dichloride,succinic chloride,1,2-bis chlorocarbonyl ethane,butanedioyl chloride,unii-gdn09v9867,1,2-ethanediylbis chloroformate
10970
butanedioyl dichloride
C(CC(=O)Cl)C(=O)Cl
543-20-4
C4H4Cl2O2
154.98
MFCD00000749
IRXBNHGNHKNOJI-UHFFFAOYSA-N
succinyl chloride,succinyl dichloride,succinic acid dichloride,succinoyl chloride,succinoyl dichloride,succinic chloride,1,2-bis chlorocarbonyl ethane,butanedioyl chloride,unii-gdn09v9867,1,2-ethanediylbis chloroformate
10970
butanedioyl dichloride
C(CC(=O)Cl)C(=O)Cl
Melting Point 16°C to 17°C
Flash Point 76°C
Boiling Point 190°C
Specific Gravity 1.407
Refractive Index 1.4700 to 1.472
Density 1.4070g/mL
Infrared Spectrum Authentic
Color Colorless to Brown
Quantity 500g
Packaging Glass bottle
Assay Percent Range ca. 95%
Linear Formula ClCOCH2CH2COCl
Beilstein 02, IV, 1921
Merck Index 15, 9004
Solubility Information Solubility in water: decomposes
Formula Weight 154.98
Physical Form Liquid
Percent Purity ≥94% (RCOCl)
Chemical Name or Material Succinyl chloride
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger