Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinylcholine chloride, 96%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
71-27-2
C14H30Cl2N2O4
361.3
YOEWQQVKRJEPAE-UHFFFAOYSA-L
succinylcholine chloride,succinyldicholine chloride,suxamethonium chloride,midarine,scoline,succinylcholine dichloride,anectine,lysthenon,succinylforte,lysthenone
22475
CHEBI:61219
trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
71-27-2
C14H30Cl2N2O4
361.3
YOEWQQVKRJEPAE-UHFFFAOYSA-L
succinylcholine chloride,succinyldicholine chloride,suxamethonium chloride,midarine,scoline,succinylcholine dichloride,anectine,lysthenon,succinylforte,lysthenone
22475
CHEBI:61219
trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]
Water 10% max.
Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Solubility Information Solubility in water: soluble.
Formula Weight 361.3
Percent Purity 96%
Chemical Name or Material Succinylcholine chloride

GHS H Statement
Toxic if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 200-747-4