Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUCLA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558787
Description
Immunogen sequence: FGGIMRCDVI AQGIVMAVKD LEIKIPVVVR LQGTRVDDAK ALIADSGLKI LACDDLDEAA RMVVKLSEIV TLAKQAHVDV KFQLP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
ATP-specific succinyl-CoA synthetase functions in the citric acid cycle (TCA), coupling the hydrolysis of succinyl-CoA to the synthesis of ATP and thus represents the only step of substrate-level phosphorylation in the TCA. The beta subunit provides nucleotide specificity of the enzyme and binds the substrate succinate, while the binding sites for coenzyme A and phosphate are found in the alpha subunit.
Specifications
SUCLA2 | |
Polyclonal | |
Unconjugated | |
SUCLA2 | |
4930547K18Rik; A-BETA; A-SCS; ATP-specific succinyl-CoA synthetase subunit beta; ATP-specific succinyl-CoA synthetase, beta subunit; mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta; MTDPS5; renal carcinoma antigen NY-REN-39; RP11-528D24.2; SCSbetaA; SCS-betaA; succinate--CoA ligase (ADP-forming); Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial; succinate-CoA ligase ADP-forming beta subunit; succinate-CoA ligase beta subunit; succinate-CoA ligase, ADP-forming, beta subunit; succinate-Coenzyme A ligase, ADP-forming, beta subunit; succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial; succinyl-CoA synthetase beta-A chain; Sucla2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20916, 8803 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2R7, Q9Z2I9 | |
SUCLA2 | |
Recombinant protein corresponding to Human SUCLA2. Recombinant protein control fragment (Product #RP-97963). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction