Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, BiotechGrade, Spectrum™

Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
57-50-1
C12H22O11
342.30
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
57-50-1
C12H22O11
342.30
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

S1699, 57-50-1, C12H22O11

Spectrum™ Sucrose, BiotechGrade - is a disaccharide composed of glucose and fructose. Sucrose is used for a variety of applications such as a supplement in plant, insect, and bacterial culture media. Spectrum offers highly pure reagents suitable for biotech research and analysis.

Packaging Poly Bottle
Appearance White, colorless
Ignition Residue 0.0005
Odor Odorless
Quantity 500 g
Specific Rotation +65.9 °
CAS Max % 1
Grade Biotech