Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, BiotechGrade, Spectrum™

Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
57-50-1
C12H22O11
342.30
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
57-50-1
C12H22O11
342.30
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

S1699, 57-50-1, C12H22O11

Spectrum™ Sucrose, BiotechGrade - is a disaccharide composed of glucose and fructose. Sucrose is used for a variety of applications such as a supplement in plant, insect, and bacterial culture media. Spectrum offers highly pure reagents suitable for biotech research and analysis.

Packaging Poly Bottle
Specific Rotation +65.9 °
Ignition Residue 0.0005
Odor Odorless
Quantity 2.5 kg
Appearance White, colorless
CAS Max % 1
Grade Biotech