Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

57-50-1
C12H22O11
342.297
CZMRCDWAGMRECN-UGDNZRGBSA-N
5988
CHEBI:17992

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
CAS 57-50-1
Molecular Formula C12H22O11
InChI Key CZMRCDWAGMRECN-UGDNZRGBSA-N
SMILES C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.297
PubChem CID 5988
Packaging Poly Bottle
ChEBI CHEBI:17992
Formula Weight 342.3
Physical Form Crystals
Specific Gravity 1L = 1.59kg
Chemical Name or Material Sucrose
Grade Baker Analyzed™ ACS Reagent
Show More Show Less