Learn More
Sucrose Gel-Loading Dye for DNA Gels, 5X (Contains 40% Sucrose/Mol. Biol.), Fisher BioReagents
$127.00

Specifications
Chemical Name or Material | Sucrose Gel-Loading Dye for DNA Gels |
---|---|
Name Note | 5X, Contains 40% Sucrose |
CAS | 57-50-1, 7732-18-5 |
Grade | Electrophoresis |
DNase | DNase free |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP655-5
![]() |
5mL | Poly Bottle |
Each for $127.00
Order in Bulk
Request bulk or custom formats |
|
Description
This dye includes sucrose instead of glycerol. Add the dye to DNA samples prior to electrophoresis on agarose gels.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Sucrose Gel-Loading Dye for DNA Gels | |
57-50-1, 7732-18-5 | |
DNase free | |
Liquid |
5X, Contains 40% Sucrose | |
Electrophoresis | |
Pass Test |
Safety and Handling

Recommended Storage : RT
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.