Sucrose, High Purity (Low Endotoxin), Multi-Compendial, N.F., J.T.Baker™

Manufacturer: J.T.Baker 400506

Packaging Poly Pail
Quantity 5kg

 View more versions of this product

Catalog No. 02-004-333

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 57-50-1
    Molecular Formula C12H22O11
    Formula Weight 342.3
    Chemical Name or Material Sucrose, High Purity (Low Endotoxin)
    Grade Multi-Compendial/NF

    Specifications

    Formula Weight 342.3
    Molecular Formula C12H22O11
    CAS 57-50-1
    Packaging Poly Pail
    Specific Gravity 1L = 1.59kg
    Quantity 5kg
    Chemical Name or Material Sucrose, High Purity (Low Endotoxin)
    Grade Multi-Compendial/NF

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit