Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sucrose, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Plastic bottle
57-50-1
C12H22O11
342.297
MFCD00006626
CZMRCDWAGMRECN-UGDNZRGBSA-N
sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
57-50-1
C12H22O11
342.297
MFCD00006626
CZMRCDWAGMRECN-UGDNZRGBSA-N
sucrose,saccharose,cane sugar,sugar,table sugar,white sugar,d-sucrose,saccharum,rohrzucker,amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
Packaging Plastic bottle
Melting Point 185°C to 187°C
Specific Gravity 1.58
Quantity 1kg
Formula Weight 342.30
Physical Form Crystalline Powder
Grade Molecular Biology
Chemical Name or Material Sucrose