Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Poly Bottle
Poly Pail
57-50-1
C12H22O11
342.297
CZMRCDWAGMRECN-UGDNZRGBSA-N
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
57-50-1
C12H22O11
342.297
CZMRCDWAGMRECN-UGDNZRGBSA-N
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Density Gradient Centrifugation and Molecular Biology Applications
Quantity 5kg
Packaging Poly Pail
Formula Weight 342.3
Specific Gravity 1L = 1.59kg
Grade Ultrapure Bioreagent
Chemical Name or Material Sucrose