Learn More
Sklar InstrumentsSupplier Diversity Partner Suction Tube Cleaning Brush, Sklar, 1.6mm x 6 in. Suction Tube Cleaning Brush For Baron and Frazier Suction Tubes, Nylon / Stainless Steel. PK/3. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101396
Suction Tube Cleaning Brush, Sklar, 1.6mm x 6 in. Suction Tube Cleaning Brush For Baron and Frazier Suction Tubes, Nylon / Stainless Steel. Sklar's Suction Tube Cleaning Brushes have been specifically designed for use with Baron and Frazier suction tubes. They are available in a number of narrow brush tips to accommodate the various size Baron and Frazier tubes. This product has a 1.6mm wide brush tip and an overall length of 6 inches. They are sold in packs of three.
Catalog No. 50-143-6972