Alfa Aesar™ Sudan III (chromatographic standard)

Manufacturer: Alfa Aesar™ A1146106

Quantity 5g

 View more versions of this product

Catalog No. AAA1146106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85-86-9
    Molecular Formula C22H16N4O
    Formula Weight 352.4
    MDL Number MFCD00003905
    Beilstein 931185
    Merck Index 14,8884
    Synonym C.I. 26100; Solvent Red 23
    Chemical Name or Material Sudan III (chromatographic standard)

    Specifications

    Formula Weight 352.4
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Warning
    Quantity 5g
    Molecular Formula C22H16N4O
    Synonym C.I. 26100; Solvent Red 23
    Melting Point ≈200°C (dec.)
    RTECS Number QK4250000
    MDL Number MFCD00003905
    CAS 85-86-9
    Beilstein 931185
    Merck Index 14,8884
    Health Hazard 2 GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Sudan III (chromatographic standard)

    GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit